Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCGTCAACAATCAGAGTGATAC[A/G]GTGTTTGCTTTTGTTAGCTTGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAMA1 - laminin subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005559.3 | 9056 | Missense Mutation | CGT,TGT | R,C 2988 | NP_005550.2 | |
XM_011525655.2 | 9056 | Missense Mutation | CGT,TGT | R,C 2998 | XP_011523957.1 | |
XM_011525656.2 | 9056 | Missense Mutation | CGT,TGT | R,C 2474 | XP_011523958.1 | |
XM_011525657.1 | 9056 | Intron | XP_011523959.1 |