Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGACCAGGGGGTCGGCGGAGCAA[C/T]AGAGACATCTGACGGTAAGCAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164360 | ||||||||||||||||||||
Literature Links: |
ATP5A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5A1 - ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001935.2 | 1100 | Silent Mutation | CTA,CTG | L,L 276 | NP_001001935.1 | |
NM_001001937.1 | 1100 | Silent Mutation | CTA,CTG | L,L 326 | NP_001001937.1 | |
NM_001257334.1 | 1100 | Silent Mutation | CTA,CTG | L,L 304 | NP_001244263.1 | |
NM_001257335.1 | 1100 | Silent Mutation | CTA,CTG | L,L 276 | NP_001244264.1 | |
NM_004046.5 | 1100 | Silent Mutation | CTA,CTG | L,L 326 | NP_004037.1 | |
XM_017025789.1 | 1100 | Silent Mutation | CTA,CTG | L,L 326 | XP_016881278.1 |