Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATTCCCTCTGCTGACAACCAAA[C/T]GTGTGTTCTGGAAGGGTGTTTTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616990 MIM: 188350 | ||||||||||||||||||||
Literature Links: |
CLUL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLUL1 - clusterin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TYMS - thymidylate synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001071.2 | 371 | Missense Mutation | CGT,TGT | R,C 78 | NP_001062.1 |
TYMSOS - TYMS opposite strand | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |