Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGATGGCTCGTTTCACTATAA[A/G]GATAAGCTGTACAGATCTGCTTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613258 MIM: 607051 | ||||||||||||||||||||
Literature Links: |
C18orf54 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C18orf54 - chromosome 18 open reading frame 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288980.1 | 257 | Silent Mutation | AAA,AAG | K,K 41 | NP_001275909.1 | |
NM_001288981.1 | 257 | Silent Mutation | AAA,AAG | K,K 41 | NP_001275910.1 | |
NM_001288982.1 | 257 | Silent Mutation | AAA,AAG | K,K 41 | NP_001275911.1 | |
NM_173529.5 | 257 | Silent Mutation | AAA,AAG | K,K 41 | NP_775800.3 | |
XM_005258201.1 | 257 | Silent Mutation | AAA,AAG | K,K 41 | XP_005258258.1 | |
XM_005258203.1 | 257 | Silent Mutation | AAA,AAG | K,K 41 | XP_005258260.1 | |
XM_017025567.1 | 257 | Silent Mutation | AAA,AAG | K,K 41 | XP_016881056.1 |
STARD6 - StAR related lipid transfer domain containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |