Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCAAGGCCCTGGGCAATCAGCAG[C/T]CTTCGTGGGATTCAGAGGATAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRP - gastrin releasing peptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012512.2 | 337 | Missense Mutation | CCT,TCT | P,S 109 | NP_001012530.1 | |
NM_001012513.2 | 337 | Missense Mutation | CCT,TCT | P,S 109 | NP_001012531.1 | |
NM_002091.4 | 337 | Missense Mutation | CCT,TCT | P,S 109 | NP_002082.2 | |
XM_011525933.2 | 337 | Missense Mutation | CCT,TCT | P,S 86 | XP_011524235.1 | |
XM_017025712.1 | 337 | Missense Mutation | CCT,TCT | P,S 109 | XP_016881201.1 | |
XM_017025713.1 | 337 | Missense Mutation | CCT,TCT | P,S 109 | XP_016881202.1 | |
XM_017025714.1 | 337 | Missense Mutation | CCT,TCT | P,S 78 | XP_016881203.1 |