Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCCTTTCCATGAGCAGCAGCAC[A/G]CCATTGCTAGCAGCTTCTCTGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600433 | ||||||||||||||||||||
Literature Links: |
CBLN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBLN2 - cerebellin 2 precursor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182511.3 | 1631 | Silent Mutation | GGC,GGT | G,G 187 | NP_872317.1 | |
XM_006722394.3 | 1631 | Silent Mutation | GGC,GGT | G,G 187 | XP_006722457.1 | |
XM_011525824.2 | 1631 | Silent Mutation | GGC,GGT | G,G 187 | XP_011524126.1 | |
XM_017025559.1 | 1631 | Silent Mutation | GGC,GGT | G,G 187 | XP_016881048.1 | |
XM_017025560.1 | 1631 | Silent Mutation | GGC,GGT | G,G 187 | XP_016881049.1 |