Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACAGCGGTCCGGTGACCAAGTC[A/G]AGGTAAGGGTGATGTCGGCTCGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF271P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF271P - zinc finger protein 271, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN30 - zinc finger and SCAN domain containing 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112734.3 | Intron | NP_001106205.1 | ||||
NM_001166012.2 | Intron | NP_001159484.1 | ||||
NM_001288711.1 | Intron | NP_001275640.1 | ||||
XM_005258183.4 | Intron | XP_005258240.1 | ||||
XM_006722371.3 | Intron | XP_006722434.1 | ||||
XM_011525789.2 | Intron | XP_011524091.1 | ||||
XM_017025515.1 | Intron | XP_016881004.1 | ||||
XM_017025516.1 | Intron | XP_016881005.1 | ||||
XM_017025517.1 | Intron | XP_016881006.1 | ||||
XM_017025518.1 | Intron | XP_016881007.1 | ||||
XM_017025519.1 | Intron | XP_016881008.1 | ||||
XM_017025520.1 | Intron | XP_016881009.1 | ||||
XM_017025521.1 | Intron | XP_016881010.1 | ||||
XM_017025522.1 | Intron | XP_016881011.1 |