Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTGGGTGCTTCCTCCAGATACT[G/T]CAGGCCAGAGGCCATCGGGAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610396 | ||||||||||||||||||||
Literature Links: |
NKPD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NKPD1 - NTPase, KAP family P-loop domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC6A - trafficking protein particle complex 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270891.1 | 356 | Missense Mutation | AAG,CAG | K,Q 111 | NP_001257820.1 | |
NM_001270892.1 | 356 | Missense Mutation | GAA,GCA | E,A 102 | NP_001257821.1 | |
NM_001270893.1 | 356 | Missense Mutation | GAA,GCA | E,A 88 | NP_001257822.1 | |
NM_024108.2 | 356 | Missense Mutation | AAG,CAG | K,Q 125 | NP_077013.1 | |
XM_017027295.1 | 356 | Missense Mutation | GAA,GCA | E,A 137 | XP_016882784.1 |