Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCTCCCGGGTGCCTGAGCCCGT[C/G]CAGAAGCGCATCGTGTTTTGGTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZSWIM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZSWIM4 - zinc finger SWIM-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023072.2 | 189 | Silent Mutation | GTC,GTG | V,V 63 | NP_075560.2 | |
XM_005260023.3 | 189 | Silent Mutation | GTC,GTG | V,V 63 | XP_005260080.1 | |
XM_017027153.1 | 189 | Silent Mutation | GTC,GTG | V,V 63 | XP_016882642.1 | |
XM_017027154.1 | 189 | Silent Mutation | GTC,GTG | V,V 63 | XP_016882643.1 | |
XM_017027155.1 | 189 | Silent Mutation | GTC,GTG | V,V 63 | XP_016882644.1 | |
XM_017027156.1 | 189 | Intron | XP_016882645.1 | |||
XM_017027157.1 | 189 | Silent Mutation | GTC,GTG | V,V 63 | XP_016882646.1 |