Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGGATGAACCCTGCTCAGAAGC[A/G]TTTATACAGGAGTATGATGTTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF471 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF471 - zinc finger protein 471 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321768.1 | 243 | Intron | NP_001308697.1 | |||
NM_020813.3 | 243 | Missense Mutation | CAT,CGT | H,R 37 | NP_065864.2 | |
XM_011527148.1 | 243 | Missense Mutation | CAT,CGT | H,R 37 | XP_011525450.1 | |
XM_017027032.1 | 243 | Intron | XP_016882521.1 |