Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTCCTGGACCCTGCCGCCTGCT[C/G]CAAACTCCACCAGCCCCGTGTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605816 | ||||||||||||||||||||
Literature Links: |
ANKRD24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD24 - ankyrin repeat domain 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EBI3 - Epstein-Barr virus induced 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005755.2 | 417 | Missense Mutation | CCA,GCA | P,A 54 | NP_005746.2 | |
XM_011527619.2 | 417 | Missense Mutation | CCA,GCA | P,A 54 | XP_011525921.1 |