Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAACTATCTGATCTCTCGGTCC[C/T]TTCCTTAACCCCATAAAAAGAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607126 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLPP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLPP2 - phospholipid phosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003712.3 | 1228 | UTR 3 | NP_003703.1 | |||
NM_177526.2 | 1228 | UTR 3 | NP_803545.1 | |||
NM_177543.2 | 1228 | UTR 3 | NP_808211.1 | |||
XM_011528396.2 | 1228 | UTR 3 | XP_011526698.1 |