Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCCCCGCAGCCCCTGCAGGAC[A/G]GTAGAAATGGGCTGGACAGAGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604814 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LILRA6 - leukocyte immunoglobulin like receptor A6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LILRB5 - leukocyte immunoglobulin like receptor B5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081442.2 | 1574 | Missense Mutation | CGT,TGT | R,C 496 | NP_001074911.1 | |
NM_001081443.2 | 1574 | Missense Mutation | CGT,TGT | R,C 396 | NP_001074912.1 | |
NM_001304457.1 | 1574 | Missense Mutation | CGT,TGT | R,C 487 | NP_001291386.1 | |
NM_006840.4 | 1574 | Missense Mutation | CGT,TGT | R,C 495 | NP_006831.1 | |
XM_011526359.2 | 1574 | Missense Mutation | CGT,TGT | R,C 496 | XP_011524661.1 | |
XM_011526360.2 | 1574 | Missense Mutation | CGT,TGT | R,C 495 | XP_011524662.1 | |
XM_011526361.2 | 1574 | Missense Mutation | CGT,TGT | R,C 479 | XP_011524663.1 | |
XM_011526362.2 | 1574 | Missense Mutation | CGT,TGT | R,C 396 | XP_011524664.1 |