Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTACATTATGAATTTTCAGAT[A/G]ATGAGTAAGGTGTGAATGTTGCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP82 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP82 - ZFP82 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321917.1 | 1831 | Missense Mutation | CAT,TAT | H,Y 525 | NP_001308846.1 | |
NM_001321918.1 | 1831 | Intron | NP_001308847.1 | |||
NM_001321919.1 | 1831 | Intron | NP_001308848.1 | |||
NM_133466.3 | 1831 | Missense Mutation | CAT,TAT | H,Y 524 | NP_597723.1 |