Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGGCTCCCTGTGTGACTGCTCC[A/G]GCAGCCCTGCCTTCTCCAAGGTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613332 | ||||||||||||||||||||
Literature Links: |
MARCH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MARCH2 - membrane associated ring-CH-type finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005415.1 | 222 | Missense Mutation | AGC,GGC | S,G 18 | NP_001005415.1 | |
NM_001005416.1 | 222 | Missense Mutation | AGC,GGC | S,G 18 | NP_001005416.1 | |
NM_016496.4 | 222 | Missense Mutation | AGC,GGC | S,G 18 | NP_057580.3 | |
XM_006722763.3 | 222 | Missense Mutation | AGC,GGC | S,G 18 | XP_006722826.1 | |
XM_011528046.2 | 222 | Missense Mutation | AGC,GGC | S,G 18 | XP_011526348.1 | |
XM_011528047.2 | 222 | Missense Mutation | AGC,GGC | S,G 18 | XP_011526349.1 | |
XM_017026853.1 | 222 | Missense Mutation | AGC,GGC | S,G 18 | XP_016882342.1 | |
XM_017026854.1 | 222 | Intron | XP_016882343.1 |