Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGAAGACAGTCAGAGAGAGACT[C/T]GCAATAGCACTGATTGCGGATGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607963 MIM: 606154 | ||||||||||||||||||||
Literature Links: |
MBD3L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBD3L1 - methyl-CpG binding domain protein 3 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145208.2 | 590 | Silent Mutation | CTC,CTT | L,L 168 | NP_660209.2 |
MUC16 - mucin 16, cell surface associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |