Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTCCCACATTCTTTACATTCAT[C/T]GGCATTCACTCCTGTGTGAGTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194557 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF20 - zinc finger protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203250.1 | 988 | Missense Mutation | AAT,GAT | N,D 252 | NP_001190179.1 | |
NM_021143.3 | 988 | Missense Mutation | AAT,GAT | N,D 255 | NP_066966.2 |
ZNF625-ZNF20 - ZNF625-ZNF20 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |