Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGCGCCTGGAGCCGCCTGAGGG[G/T]CCTGGGACCTCTAGAGCCCTGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607769 MIM: 611048 MIM: 602309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHA4 - pleckstrin homology domain containing A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R15A - protein phosphatase 1 regulatory subunit 15A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014330.3 | 382 | Missense Mutation | GGC,GTC | G,V 38 | NP_055145.3 |
TULP2 - tubby like protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |