Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGACATGGCACCCTGGGCCTGGG[A/G]GTCGGCGTCAGTGGCCTCCTCGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610336 MIM: 605217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD4C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2CD4C - C2 calcium dependent domain containing 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136263.1 | 551 | Missense Mutation | CCC,TCC | P,S 130 | NP_001129735.1 | |
XM_011527694.1 | 551 | Missense Mutation | CCC,TCC | P,S 130 | XP_011525996.1 |
SHC2 - SHC adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |