Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGACTCACATCAAAGAACTGTCC[A/G]GGACCAAATGGGTTGTGGGTGATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602518 | ||||||||||||||||||||
Literature Links: |
LGALS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LGALS4 - galectin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006149.3 | 1429 | Silent Mutation | CCC,CCT | P,P 270 | NP_006140.1 | |
XM_011526973.1 | 1429 | Silent Mutation | CCC,CCT | P,P 257 | XP_011525275.1 | |
XM_011526974.1 | 1429 | Silent Mutation | CCC,CCT | P,P 139 | XP_011525276.1 |