Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCCAGGGGCTGCACGGATATCA[C/T]ATGCTGTGTGTTCCTGCTCCTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606106 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC44A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC44A2 - solute carrier family 44 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145056.1 | 243 | Missense Mutation | ACA,ATA | T,I 33 | NP_001138528.1 | |
NM_020428.3 | 243 | Missense Mutation | ACA,ATA | T,I 35 | NP_065161.3 | |
XM_005259997.1 | 243 | Missense Mutation | ACA,ATA | T,I 35 | XP_005260054.1 | |
XM_005259999.1 | 243 | Missense Mutation | ACA,ATA | T,I 33 | XP_005260056.1 |