Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTATTAAGTTACATTATGAATTC[A/G]CTGATGGTAAGTAAGATGTGAATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZFP30 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFP30 - ZFP30 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320666.1 | 1862 | Nonsense Mutation | CGA,TGA | R,* 514 | NP_001307595.1 | |
NM_001320667.1 | 1862 | Nonsense Mutation | CGA,TGA | R,* 514 | NP_001307596.1 | |
NM_001320668.1 | 1862 | Nonsense Mutation | CGA,TGA | R,* 514 | NP_001307597.1 | |
NM_001320669.1 | 1862 | Nonsense Mutation | CGA,TGA | R,* 514 | NP_001307598.1 | |
NM_001320670.1 | 1862 | Nonsense Mutation | CGA,TGA | R,* 513 | NP_001307599.1 | |
NM_001320671.1 | 1862 | Nonsense Mutation | CGA,TGA | R,* 513 | NP_001307600.1 | |
NM_014898.2 | 1862 | Nonsense Mutation | CGA,TGA | R,* 514 | NP_055713.1 |