Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATCCAGAAGGGCCCTGCAGGCG[A/G]AGGGACATCCAGGGCCTGGAAACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605631 | ||||||||||||||||||||
Literature Links: |
NAPSA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAPSA - napsin A aspartic peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004851.2 | 1628 | Missense Mutation | CCG,TCG | P,S 365 | NP_004842.1 | |
XM_011527540.1 | 1628 | Missense Mutation | CCG,TCG | P,S 365 | XP_011525842.1 | |
XM_017027512.1 | 1628 | Missense Mutation | CCG,TCG | P,S 356 | XP_016883001.1 |