Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTTCCACCGAGCAACAGCTTCA[A/G]GTCATCTTTGAGGTGCTGGAGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602717 | ||||||||||||||||||||
Literature Links: |
GRIN2D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRIN2D - glutamate ionotropic receptor NMDA type subunit 2D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000836.2 | 607 | Silent Mutation | CAA,CAG | Q,Q 173 | NP_000827.2 | |
XM_011526872.1 | 607 | Silent Mutation | CAA,CAG | Q,Q 173 | XP_011525174.1 |