Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCATCAGAGGAATCCGCCCCCA[A/C]GACGTCCCCAGAGTCTGTGTCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109091 MIM: 605162 MIM: 600061 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALR - calreticulin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GADD45GIP1 - GADD45G interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6515 - microRNA 6515 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAD23A - RAD23 homolog A, nucleotide excision repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270362.1 | 527 | Missense Mutation | AAG,ACG | K,T 131 | NP_001257291.1 | |
NM_001270363.1 | 527 | Missense Mutation | AAG,ACG | K,T 131 | NP_001257292.1 | |
NM_005053.3 | 527 | Missense Mutation | AAG,ACG | K,T 131 | NP_005044.1 |