Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTTCCTTTGCCAGGTCATGCG[A/C]GGGGCTGCTGGCCACCCCATGCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609742 MIM: 616659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL4I1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL4I1 - interleukin 4 induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258017.1 | 2241 | Missense Mutation | GCG,TCG | A,S 560 | NP_001244946.1 | |
NM_001258018.1 | 2241 | Missense Mutation | GCG,TCG | A,S 560 | NP_001244947.1 | |
NM_152899.1 | 2241 | Missense Mutation | GCG,TCG | A,S 538 | NP_690863.1 | |
NM_172374.2 | 2241 | Missense Mutation | GCG,TCG | A,S 560 | NP_758962.1 |
MIR4750 - microRNA 4750 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D17 - TBC1 domain family member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |