Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCACCTGGTGAAGCGGACCTT[A/G]CAGATGTTGCACTCGTAGGGCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBTB7A - zinc finger and BTB domain containing 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317990.1 | 2446 | Silent Mutation | TGC,TGT | C,C 415 | NP_001304919.1 | |
NM_015898.3 | 2446 | Silent Mutation | TGC,TGT | C,C 415 | NP_056982.1 | |
XM_005259570.4 | 2446 | Silent Mutation | TGC,TGT | C,C 465 | XP_005259627.3 | |
XM_005259571.4 | 2446 | Silent Mutation | TGC,TGT | C,C 415 | XP_005259628.1 |