Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACCTGTCCATTTCAGGCATCAA[C/T]ATTTATGAGCCAGCACCCCCTACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615901 | ||||||||||||||||||||
Literature Links: |
NCCRP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCCRP1 - non-specific cytotoxic cell receptor protein 1 homolog (zebrafish) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001414.1 | 364 | Silent Mutation | AAC,AAT | N,N 115 | NP_001001414.1 | |
XM_011526906.2 | 364 | Silent Mutation | AAC,AAT | N,N 115 | XP_011525208.1 |
SYCN - syncollin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |