Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAAGGGAGGGCAGGGATCGCC[A/G]AGACCACCACCGGGCTCCCAGCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615475 | ||||||||||||||||||||
Literature Links: |
DHX34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX34 - DEAH-box helicase 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014681.5 | 336 | Missense Mutation | CAA,CGA | Q,R 13 | NP_055496.2 | |
XM_005259443.3 | 336 | Missense Mutation | CAA,CGA | Q,R 13 | XP_005259500.1 | |
XM_011527550.2 | 336 | Missense Mutation | CAA,CGA | Q,R 13 | XP_011525852.1 | |
XM_011527551.2 | 336 | Missense Mutation | CAA,CGA | Q,R 13 | XP_011525853.1 | |
XM_017027524.1 | 336 | UTR 5 | XP_016883013.1 |