Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTCTCCTGTGTGAACTCTCTCA[C/T]GTACAATGAGGTCAAATTTCCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606956 | ||||||||||||||||||||
Literature Links: |
ZNF256 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF256 - zinc finger protein 256 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005773.2 | 1593 | Missense Mutation | CAT,CGT | H,R 453 | NP_005764.2 | |
XM_017026138.1 | 1593 | Missense Mutation | CAT,CGT | H,R 300 | XP_016881627.1 |
ZNF418 - zinc finger protein 418 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |