Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGATCTGACGAGCTTTTCTCTA[C/T]TTGTGTCACTAACGGACCGTTTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610391 MIM: 600693 | ||||||||||||||||||||
Literature Links: |
MIR3187 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR3187 - microRNA 3187 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4745 - microRNA 4745 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLPPR3 - phospholipid phosphatase related 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTBP1 - polypyrimidine tract binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002819.4 | 674 | Missense Mutation | ACT,ATT | T,I 22 | NP_002810.1 | |
NM_031990.3 | 674 | Missense Mutation | ACT,ATT | T,I 22 | NP_114367.1 | |
NM_031991.3 | 674 | Missense Mutation | ACT,ATT | T,I 22 | NP_114368.1 | |
XM_005259597.2 | 674 | Missense Mutation | ACT,ATT | T,I 24 | XP_005259654.1 | |
XM_005259598.2 | 674 | Missense Mutation | ACT,ATT | T,I 24 | XP_005259655.1 |