Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTAGAACATTGGAACCTCTCCC[C/T]TTTAGGATGAAACATTGGAAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 128260 MIM: 613513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNRPE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNRPE - small nuclear ribonucleoprotein polypeptide E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304464.1 | Intron | NP_001291393.1 | ||||
NM_003094.3 | Intron | NP_003085.1 |
ZC3H11A - zinc finger CCCH-type containing 11A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |