Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCTGGTGCAGGCTCTTGGGGCC[G/T]GCGGCGGCGACGAAGGCTGCGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607348 MIM: 606162 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HES5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HES5 - hes family bHLH transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010926.3 | 318 | Silent Mutation | GCA,GCC | A,A 79 | NP_001010926.1 | |
XM_005244751.4 | 318 | Silent Mutation | GCA,GCC | A,A 111 | XP_005244808.1 |
PANK4 - pantothenate kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |