Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAACTGCCAAAGTCATCACTAT[A/G]CAGCCGAGGTATGGGTGGTAACCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611578 | ||||||||||||||||||||
Literature Links: |
FRRS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRRS1 - ferric chelate reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013660.2 | 1482 | Silent Mutation | TGC,TGT | C,C 451 | NP_001013682.2 | |
XM_005270861.3 | 1482 | Silent Mutation | TGC,TGT | C,C 451 | XP_005270918.1 | |
XM_011541451.2 | 1482 | Silent Mutation | TGC,TGT | C,C 451 | XP_011539753.1 | |
XM_011541452.1 | 1482 | Silent Mutation | TGC,TGT | C,C 451 | XP_011539754.1 | |
XM_017001271.1 | 1482 | Silent Mutation | TGC,TGT | C,C 451 | XP_016856760.1 |