Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAGTTTTAATCTAGAGATTGTC[C/T]CCTGCTTGTTCCTGTCCTGTCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602024 MIM: 610692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCNKA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLCNKA - chloride voltage-gated channel Ka | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042704.1 | Intron | NP_001036169.1 | ||||
NM_001257139.1 | Intron | NP_001244068.1 | ||||
NM_004070.3 | Intron | NP_004061.3 |
HSPB7 - heat shock protein family B (small) member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |