Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGTTTCAATCCACAGACATTT[A/G]ATTACAAAGCAGTTAAACAAGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEX35 - testis expressed 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170722.1 | 209 | Missense Mutation | AAT,GAT | N,D 41 | NP_001164193.1 | |
NM_001170723.1 | 209 | Missense Mutation | AAT,GAT | N,D 33 | NP_001164194.1 | |
NM_001170724.1 | 209 | Missense Mutation | AAT,GAT | N,D 33 | NP_001164195.1 | |
NM_032126.4 | 209 | Missense Mutation | AAT,GAT | N,D 33 | NP_115502.2 |