Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACTCAGTCAGTGCTATATTGTG[C/T]CTCCATTTTACTTCCAGGACCTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HP1BP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HP1BP3 - heterochromatin protein 1 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016287.3 | 2650 | UTR 3 | NP_057371.2 | |||
XM_005245875.4 | 2650 | UTR 3 | XP_005245932.1 | |||
XM_005245876.3 | 2650 | UTR 3 | XP_005245933.1 | |||
XM_005245877.4 | 2650 | UTR 3 | XP_005245934.1 | |||
XM_005245878.4 | 2650 | UTR 3 | XP_005245935.1 | |||
XM_005245879.4 | 2650 | UTR 3 | XP_005245936.1 | |||
XM_011541532.1 | 2650 | UTR 3 | XP_011539834.1 | |||
XM_011541533.1 | 2650 | UTR 3 | XP_011539835.1 | |||
XM_011541534.2 | 2650 | UTR 3 | XP_011539836.1 | |||
XM_017001393.1 | 2650 | UTR 3 | XP_016856882.1 | |||
XM_017001394.1 | 2650 | UTR 3 | XP_016856883.1 |