Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGAGCCTTTCAGCCACAGAAA[C/T]GATTGACATGCTTCTCTCCCCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150330 MIM: 611007 MIM: 612942 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMNA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMNA - lamin A/C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEX3A - mex-3 RNA binding family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093725.1 | 2173 | UTR 3 | NP_001087194.1 |
RAB25 - RAB25, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |