Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTTTCTTCAATTAAAAAACAA[C/T]TTCAGCTCTTCTATTATAAACATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRB1 - crumbs 1, cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193640.1 | Intron | NP_001180569.1 | ||||
NM_001257965.1 | Intron | NP_001244894.1 | ||||
NM_001257966.1 | Intron | NP_001244895.1 | ||||
NM_201253.2 | Intron | NP_957705.1 | ||||
XM_011509365.2 | Intron | XP_011507667.1 | ||||
XM_011509367.1 | Intron | XP_011507669.1 | ||||
XM_011509369.2 | Intron | XP_011507671.1 | ||||
XM_017000851.1 | Intron | XP_016856340.1 | ||||
XM_017000852.1 | Intron | XP_016856341.1 |