Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCAAGAGAATTCCTAACCAGAA[C/T]CCCAGGCCCCCAACTCCTTAACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPCAL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPCAL4 - hippocalcin like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282396.1 | 1299 | UTR 3 | NP_001269325.1 | |||
NM_001282397.1 | 1299 | UTR 3 | NP_001269326.1 | |||
NM_016257.3 | 1299 | UTR 3 | NP_057341.1 | |||
XM_017001447.1 | 1299 | UTR 3 | XP_016856936.1 |