Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGCAGCTTTAGTGACTGGCTC[A/G]GTTATACTAGAGCATTGGGATTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600716 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP4B1-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP4B1-AS1 - AP4B1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BCL2L15 - BCL2 like 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010922.2 | 4597 | UTR 3 | NP_001010922.1 |
PTPN22 - protein tyrosine phosphatase, non-receptor type 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |