Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACATGGCTCTGGGCACTTTGGC[A/G]TGGGGCACACAGGAGGTGGCTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182269 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRR2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPRR2C - small proline rich protein 2C (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPRR2G - small proline rich protein 2G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014291.3 | 103 | Missense Mutation | ACG,ATG | T,M 20 | NP_001014313.1 | |
XM_017002177.1 | 103 | Missense Mutation | ACG,ATG | T,M 20 | XP_016857666.1 | |
XM_017002178.1 | 103 | Missense Mutation | ACG,ATG | T,M 20 | XP_016857667.1 |