Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGTTGGTTGTTCCTAGAGCTTT[C/T]GCTGCATTGGTTTTTGCTATTTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSRP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RSRP1 - arginine and serine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321772.1 | 803 | Silent Mutation | GCA,GCG | A,A 187 | NP_001308701.1 | |
NM_020317.4 | 803 | Silent Mutation | GCA,GCG | A,A 187 | NP_064713.3 | |
XM_011541797.1 | 803 | Silent Mutation | GCA,GCG | A,A 187 | XP_011540099.1 |