Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTGCATTCTCCAGGAAAGCTC[C/T]GGAAGTCCCACATCCCTGGAGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGFN1 - immunoglobulin-like and fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164586.1 | 191 | Missense Mutation | CGG,TGG | R,W 6 | NP_001158058.1 | |
XM_005245580.3 | 191 | Missense Mutation | CGG,TGG | R,W 6 | XP_005245637.1 | |
XM_006711618.3 | 191 | Missense Mutation | CGG,TGG | R,W 6 | XP_006711681.1 | |
XM_011510120.2 | 191 | Missense Mutation | CGG,TGG | R,W 6 | XP_011508422.1 | |
XM_011510121.2 | 191 | Missense Mutation | CGG,TGG | R,W 6 | XP_011508423.1 | |
XM_017002787.1 | 191 | Missense Mutation | CGG,TGG | R,W 6 | XP_016858276.1 |