Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGGCCCTGGTGGGCAACACGCTG[A/G]GTAGGTCCAGGGCTTGCCCGGCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602392 | ||||||||||||||||||||
Literature Links: |
HCRTR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCRTR1 - hypocretin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001525.2 | 1075 | Missense Mutation | ATC,GTC | I,V 67 | NP_001516.2 | |
XM_017001105.1 | 1075 | Missense Mutation | ATC,GTC | I,V 67 | XP_016856594.1 | |
XM_017001106.1 | 1075 | Missense Mutation | ATC,GTC | I,V 67 | XP_016856595.1 | |
XM_017001107.1 | 1075 | Missense Mutation | ATC,GTC | I,V 67 | XP_016856596.1 |