Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGGTATTAAAGGAATGCACAAA[A/C]TAGTTTACAACCAACCCTAAACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603181 MIM: 607007 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ILF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ILF2 - interleukin enhancer binding factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267809.1 | Intron | NP_001254738.1 | ||||
NM_004515.3 | Intron | NP_004506.2 |
SNAPIN - SNAP associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |