Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTGAGCGCCTCTTAAAAGACTA[C/T]GAAGATAAAATTCGGGAGGAGTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615164 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKIRIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKIRIN1 - akirin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136275.1 | 581 | Intron | NP_001129747.1 | |||
NM_024595.2 | 581 | Silent Mutation | TAC,TAT | Y,Y 148 | NP_078871.1 |