Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCAGAATTGGACTCCCCCTCACT[C/T]GCAGTAGAGCCCGTGGACCGTGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602913 MIM: 611713 | ||||||||||||||||||||
Literature Links: |
BTBD19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTBD19 - BTB domain containing 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLK3 - polo like kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004073.3 | 804 | Intron | NP_004064.2 |
TCTEX1D4 - Tctex1 domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013632.3 | 804 | Missense Mutation | AAG,GAG | K,E 221 | NP_001013654.1 | |
XM_006710614.3 | 804 | Missense Mutation | AAG,GAG | K,E 279 | XP_006710677.1 | |
XM_011541375.2 | 804 | Missense Mutation | AAG,GAG | K,E 221 | XP_011539677.1 | |
XM_011541376.2 | 804 | Missense Mutation | AAG,GAG | K,E 221 | XP_011539678.1 | |
XM_017001187.1 | 804 | Missense Mutation | AAG,GAG | K,E 262 | XP_016856676.1 |