Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCCAGCTCTTTGCGGCTCAATC[A/G]AGCAGGGTTCTGGGCTTTAGGTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605554 MIM: 600684 | ||||||||||||||||||||
Literature Links: |
CD244 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD244 - CD244 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166663.1 | 1294 | Nonsense Mutation | CGA,TGA | R,* 357 | NP_001160135.1 | |
NM_001166664.1 | 1294 | Nonsense Mutation | CGA,TGA | R,* 260 | NP_001160136.1 | |
NM_016382.3 | 1294 | Nonsense Mutation | CGA,TGA | R,* 352 | NP_057466.1 | |
XM_011509621.2 | 1294 | Intron | XP_011507923.1 | |||
XM_011509622.2 | 1294 | Intron | XP_011507924.1 | |||
XM_011509623.2 | 1294 | Nonsense Mutation | CGA,TGA | R,* 154 | XP_011507925.1 |
LY9 - lymphocyte antigen 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |