Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGACAGAGGCAGGCAAGTGAGC[A/G]CACATACTCCACAAAGTCCACCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176992 MIM: 114210 MIM: 176991 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928034 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928034 - uncharacterized LOC101928034 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S100A3 - S100 calcium binding protein A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002960.1 | 312 | Missense Mutation | CGC,TGC | R,C 77 | NP_002951.1 |
S100A4 - S100 calcium binding protein A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S100A5 - S100 calcium binding protein A5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |